missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-methylphenol-2,6-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-3-methylphenol 2,6-d2 |
| CAS | 93951-72-5 |
| Molecular Formula | C7 2H2 H5 Cl O |
| Quantity | 0.1 g |
| Synonym | Phen-2,6-d2-ol, 4-chloro-3-methyl- (9 CI), 4-Chloro-3-methylphenol (ring-2,6-D2), 4-Chloro-3-methylphenol-2,6-D2, 4-Chloro-2,6-dideuterio-3-methylphenol |
| InChI Formula | InChI=1 S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9 H,1H3/i2D,4 D |
| SMILES | [2 H]c1cc(Cl)c(C)c([2 H])c1O |
| IUPAC Name | 4-chloro-2,6-dideuterio-3-methylphenol |
| Molecular Weight (g/mol) | 144.6 |
| Formula Weight | 144.0311 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction