missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide-2,3,3-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Acrylamide-d3 |
| CAS | 122775-19-3 |
| Molecular Formula | C3 2H3 H2 N O |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1D2,2 D |
| SMILES | [2 H]C(=C([2 H])C(=O)N)[2 H] |
| IUPAC Name | 2,3,3-trideuterioprop-2-enamide |
| Molecular Weight (g/mol) | 74.1 |
| Formula Weight | 74.0559 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction