missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine-2,2,3,3-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | beta-Alanine-2,2,3,3-D4 |
| CAS | 116173-67-2 |
| Molecular Formula | C3 D4 H3 N O2 |
| Quantity | 0.1 g |
| Synonym | β-Alanine-2,2,3,3-d4 (9 CI, ACI), β-Alanine-d4, β-Alanine-2,2,3,3-D4, 3-Aminopropanoic-2,2,3,3-d4 acid, 3-Aminopropionic-d4 acid |
| InChI Formula | InChI=1 S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 3-amino-2,2,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 93.118 |
| Formula Weight | 93.073 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction