missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyric Anhydride-3,3,3',3',4,4,4,4',4',4'-d10, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Butyric Anhydride-3,3,3',3',4,4,4,4',4',4' D10 |
| CAS | 1398065-70-7 |
| Molecular Formula | C8 D10 H4 O3 |
| Quantity | 0.1 g |
| Synonym | Butyric anhydride-3,3,3',3',4,4,4,4',4',4' D10, 3,3,4,4,4-Pentadeuteriobutanoyl 3,3,4,4,4-pentadeuteriobutanoate, Butanoic-3,3,4,4,4-d5 acid 1,1'-anhydride, Butanoic acid anhydride-d10, Butyric anhydride-d10, Butanoic-3,3,4,4,4-d5 anhydride, Butanoyl-3,3,4,4,4-d5 anhydride, Butanoyl-3,3,4,4,4-d5 butanoate-3,3,4,4,4-d5, Butyric acid anhydride-d10, Butyryl anhydride-d10 |
| InChI Formula | InChI=1 S/C8H14O3/c1-3-5-7(9)11-8(10)6-4-2/h3-6H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CC(=O)OC(=O)CC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,4,4,4-pentadeuteriobutanoyl 3,3,4,4,4-pentadeuteriobutanoate |
| Molecular Weight (g/mol) | 168.257 |
| Formula Weight | 168.157 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction