missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Tri(octanoate-d15), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Glyceryl Trioctanoate-D45 |
| CAS | 352431-24-4 |
| Molecular Formula | C27 D45 H5 O6 |
| Quantity | 0.1 g |
| Synonym | Octanoic-d15 acid, 1,2,3-propanetriyl ester (9 CI), Octanoic-d15 acid 1,2,3-propanetriyl ester, 1,2,3-Propanetriol tris(octanoate-2,2,3,3,4,4,5,6,6,7,7,8,8,8-d15), Glyceryl Tri(octanoate-d15), Glyceryl Trioctanoate-d45, Glyceryl Trioctanoate-D45 |
| InChI Formula | InChI=1 S/C27H50O6/c1-4-7-10-13-16-19-25(28)31-22-24(33-27(30)21-18-15-12-9-6-3)23-32-26(29)20-17-14-11-8-5-2/h24H,4-23H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OCC(COC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])OC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis(2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoyloxy)propyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoate |
| Molecular Weight (g/mol) | 515.9596 |
| Formula Weight | 515.6432 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction