missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-2,4,5,6,7-d5-3-acetic Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1 H-Indole-3-acetic acid indole-2,4,5,6,7-d5 |
| CAS | 76937-78-5 |
| Molecular Formula | C10 2H5 H4 N O2 |
| Quantity | 0.1 g |
| Synonym | 1 H-Indole-3-acetic acid D5 (indole-2,4,5,6,7-D5), 1 H-Indole-2,4,5,6,7-d5-3-acetic acid, 2-(2,4,5,6,7-Pentadeuterio-1 H-indol-3-yl)acetic acid, 2-(3-Indolyl-2,4,5,6,7-d5)acetic acid, 3-Indolylacetic acid-d5 |
| InChI Formula | InChI=1 S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11 H,5H2,(H,12,13)/i1D,2 D,3 D,4 D,6 D |
| SMILES | [2 H]c1[nH]c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1CC(=O)O |
| IUPAC Name | 2-(2,4,5,6,7-pentadeuterio-1 H-indol-3-yl)acetic acid |
| Molecular Weight (g/mol) | 180.21 |
| Formula Weight | 180.0947 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction