Learn More
L-Aspartic-2,3,3-d3 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | L-Aspartic Acid-d3 |
| CAS | 3842-25-9 |
| Molecular Formula | C4 2H3 H4 N O4 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1/i1D2,2 D |
| SMILES | [2 H]C([2 H])(C(=O)O)[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,3-trideuteriobutanedioic acid |
| Molecular Weight (g/mol) | 136.12 |
| Formula Weight | 136.0563 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |