missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Hydroxyphenyl-2,3,5,6-d4)acetamide-2,2,2-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Paracetamol-D7 |
| CAS | 1219798-53-4 |
| Molecular Formula | C8 D7 H2 N O2 |
| Quantity | 0.1 g |
| Synonym | Acetamide-2,2,2-d3, N-(4-hydroxyphenyl-2,3,5,6-d4)-, Paracetamol-D4 (acetyl-D3,phenyl-D4), Paracetamol-D7, Paracetamol-d7, Acetaminophen-d7, 4-Hydroxy-2,3,5,6-tetradeutero(trideuteroacet)anilide, N-(4-Hydroxyphenyl-2,3,5,6-d4)acetamide-2,2,2-d3 |
| InChI Formula | InChI=1 S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11 H,1H3,(H,9,10)/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(NC(=O)C([2 H])([2 H])[2 H])c([2 H])c([2 H])c1O |
| IUPAC Name | 2,2,2-trideuterio-N-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetamide |
| Molecular Weight (g/mol) | 158.206 |
| Formula Weight | 158.107 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction