Learn More
N-Acetyl-d3-glycine-2,2-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-Acetylglycine-D5 (N-acetyl,2,2-D5) |
| CAS | 1219805-82-9 |
| Molecular Formula | C4 D5 H2 N O3 |
| Quantity | 0.1 g |
| Synonym | N-Acetylglycine-d5, N-Acetyl-d3-glycine-2,2-d2, N-Acetylglycine-D5 (N-acetyl,2,2-D5), N-Acetylglycine-d5, 2-(Acet-d3-amido)acetic-2,2-d2 acid |
| InChI Formula | InChI=1 S/C4H7NO3/c1-3(6)5-2-4(7)8/h2H2,1H3,(H,5,6)(H,7,8)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)NC([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-[(2,2,2-trideuterioacetyl)amino]acetic acid |
| Molecular Weight (g/mol) | 122.134 |
| Formula Weight | 122.074 g/mol |
| Show More |