missing translation for 'onlineSavingsMsg'
Learn More

n-Butyraldehyde-d8, CDN

Product Code. 30316541
missing translation for 'orderingAttributeHoverText'
Quantity:
0.05 g
0.1 g
missing translation for 'unitSize'
0.05g
0.1g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30316541

missing translation for 'mfr': CDN CDND53150.1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Butanal-d8
CAS 84965-36-6
Molecular Formula C4 2H8 O
Quantity 0.1 g
Synonym Butanal-d8
InChI Formula InChI=1 S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2,4 D
SMILES [2 H]C(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,2,2,3,3,4,4,4-octadeuteriobutan-1-one
Molecular Weight (g/mol) 80.16
Formula Weight 80.1077 g/mol
Percent Purity 99 atom % D, min 96% Chemical Purity
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.