Learn More
N,N-Dimethyl-d6-glycine HCl, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl-d6-glycine hydrochloride |
| CAS | 347840-03-3 |
| Molecular Formula | C4 2H6 H3 N O2 . Cl H |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C4H9NO2.ClH/c1-5(2)3-4(6)7;/h3H2,1-2H3,(H,6,7);1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(CC(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[bis(trideuteriomethyl)amino]acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 145.62 |
| Formula Weight | 145.0777 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |