missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosobenzylmethyl-d3-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-N-methylbenzylamine-D3 (N-methyl-D3) |
| CAS | 62845-80-1 |
| Molecular Formula | C8H7D3N2O |
| Quantity | 0.1 g |
| Synonym | Benzenemethanamine, N-(methyl-d3)-N-nitroso- (9 CI), N-Nitroso-N-methylbenzylamine-D3 (N-methyl-D3), N-(Methyl-d3)-N-nitrosobenzenemethanamine, N-Nitroso-N-methylbenzylamine-d3, N-Nitroso-N-(methyl-d3)benzylamine |
| InChI Formula | InChI=1 S/C8H10N2O/c1-10(9-11)7-8-5-3-2-4-6-8/h2-6 H,7H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N(N=O)CC1=CC=CC=C1 |
| IUPAC Name | N-benzyl-N-(methyl-d3)nitrous amide |
| Molecular Weight (g/mol) | 153.2 |
| Formula Weight | 153.1 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction