missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetyl CoA synthetase Antibody (011), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89801APC
This item is not returnable.
View return policy
Description
Acetoacetyl CoA synthetase Monoclonal antibody specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for ELISA
Specifications
| Acetoacetyl CoA synthetase | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 65985 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 011 | |
| ELISA | |
| acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Acetoacetyl CoA synthetase (Accession#: NP_076417.2; Met1-Phe672). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction