missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-2 Antibody (994930) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6628JF585
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Dkk-2 Monoclonal antibody specifically detects Dkk-2 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Tekniske data
| Dkk-2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, dickkopf-2, dickkopf-related protein 2, DKK-2, hDkk-2 | |
| Chinese Hamster Ovary cell line, CHO-derived human Dkk-2, Met1-Ile259, Accession # NP_055236 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 994930 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Stem Cells | |
| 27123 | |
| Store at 4°C in the dark. | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion