Learn More
Jak3 Antibody (452524) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB46991JF669
Product Code. 30174253
  • 605.00€ / 0.10mL
Description
Jak3 Monoclonal antibody specifically detects Jak3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Jak3
Monoclonal
Janelia Fluor 669
EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte)
E. coli-derived recombinant human Jak3, Gly46-Thr209, Accession # P52333
RUO
Primary
Human
Liquid
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
452524
50mM Sodium Borate
Mouse
0.1 mL
Cell Biology, Protein Kinase, Signal Transduction, Tyrosine Kinases
3718
Store at 4°C in the dark.
IgG2b