missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXR/NR1I2 Antibody (V11P4G11*E7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50519MFV500
This item is not returnable.
View return policy
Description
PXR/NR1I2 Monoclonal antibody specifically detects PXR/NR1I2 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| PXR/NR1I2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Zebrafish | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| V11P4G11*E7 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR | |
| Ovalbumin-conjugated synthetic peptide EMRSMNEEYT (C-terminal sequence) | |
| 0.1 mL | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| 8856 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction