missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QA1b Antibody (6A8.6F10.1A6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-26649IR
This item is not returnable.
View return policy
Description
QA1b Monoclonal antibody specifically detects QA1b in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization
Specifications
| QA1b | |
| Monoclonal | |
| DyLight 755 | |
| H2-Q1, H2-T23 | |
| Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells.The antibody reacts with the Qa-1b Class Ib molecule. | |
| 0.1 mL | |
| Signal Transduction | |
| 15040 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization | |
| 6A8.6F10.1A6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction