Learn More
1,2-Dibromobenzene-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dibromobenzene-D4 |
| CAS | 1643577-34-7 |
| Molecular Formula | C6 D4 Br2 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,3,4-d4, 5,6-dibromo- (ACI), 5,6-Dibromobenzene-1,2,3,4-d4 (ACI), 1,2-Dibromobenzene-d4, 1,2-Dibromobenzene-D4 |
| InChI Formula | InChI=1 S/C6H4Br2/c7-5-3-1-2-4-6(5)8/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Br)c(Br)c1[2 H] |
| IUPAC Name | 1,2-dibromo-3,4,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 239.9286 |
| Formula Weight | 237.8931 g/mol |
| Show More |