Learn More
1-Chlorohexane-d13, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Chlorohexane-D13 |
| CAS | 1219798-45-4 |
| Molecular Formula | C6 D13 Cl |
| Quantity | 0.25 g |
| Synonym | 6-Chlorohexane-1,1,1,2,2,3,3,4,4,5,5,6,6-d13, 1-Chlorohexane-d13, 1-Chlorohexane-D13, Hexyl-d13 chloride, n-Hexyl chloride-d13 |
| InChI Formula | InChI=1 S/C6H13Cl/c1-2-3-4-5-6-7/h2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 1-chloro-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexane |
| Molecular Weight (g/mol) | 133.7005 |
| Formula Weight | 133.1522 g/mol |
| Show More |