missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexane-d13-thiol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Hexanethiol-D13 |
| CAS | 1142922-50-6 |
| Molecular Formula | C6 D13 H S |
| Quantity | 0.25 g |
| Synonym | 1-Hexane-1,1,2,2,3,3,4,4,5,5,6,6,6-d13-thiol (ACI), 1-Hexane-d13-thiol, 1-Hexanethiol-d13, 1-Hexanethiol-D13, Hexyl-d13 Mercaptan |
| InChI Formula | InChI=1 S/C6H14S/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexane-1-thiol |
| Molecular Weight (g/mol) | 131.3205 |
| Formula Weight | 131.1632 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction