Learn More
2,4-Dinitrotoluene-alpha,alpha,alpha-d3, CDN
Product Code. 30320002
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320002
Brand: CDN CDND50930.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Dinitrotoluene methyl-d3
CAS 70786-68-4
Molecular Formula C7 2H3 H3 N2 O4
Quantity 0.25 g
Synonym Benzene, 1-(methyl-d3)-2,4-dinitro- (9 CI), 2,4-Dinitrotoluene (methyl-D3), 1-Trideuteromethyl-2,4-dinitrobenzene
InChI Formula InChI=1 S/C7H6N2O4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4 H,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])c1ccc(cc1[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2,4-dinitro-1-(trideuteriomethyl)benzene
Molecular Weight (g/mol) 185.15
Formula Weight 185.0516 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity