Learn More
3,4-Difluorobenzoic-d3 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3,4-Difluorobenzoic Acid-D3 |
| CAS | 1219798-70-5 |
| Molecular Formula | C7 D3 H F2 O2 |
| Quantity | 0.25 g |
| Synonym | 3,4-Difluorobenzoic-d3 Acid, 3,4-Difluorobenzoic-2,5,6-d3 acid, 4,5-Difluorobenzoic-2,3,6-d3 acid, 3,4-Difluorobenzoic acid-d3, 3,4-Difluorobenzoic Acid-D3 |
| InChI Formula | InChI=1 S/C7H4F2O2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3 H,(H,10,11)/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)O)c([2 H])c(F)c1F |
| IUPAC Name | 2,3,6-trideuterio-4,5-difluorobenzoic acid |
| Molecular Weight (g/mol) | 161.1208 |
| Formula Weight | 161.0368 g/mol |
| Show More |