missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Phenyl-d5-alanine-2,3,3-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | L-Phenylalanine-d8 |
| CAS | 29909-00-0 |
| Molecular Formula | C9 D8 H3 N O2 |
| Quantity | 0.25 g |
| Synonym | (2 S)-2-Amino-3-phenylpropanoic Acid-d8, (S)-(-)-Phenylalanine-d8, (S)-α-Amino-β-phenylpropionic Acid-d8, (S)-α-Aminohydrocinnamic Acid-d8, (S)-α-Amino-benzenepropanoic Acid-d8, DL-Phenylalanine-D8, DL-Phenyl-d5-alanine-2,3,3-d3 |
| InChI Formula | InChI=1 S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8 H,6,10H2,(H,11,12)/i1D,2 D,3 D,4 D,5 D,6D2,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3-trideuterio-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 173.24 |
| Formula Weight | 173.1292 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction