missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-alpha,alpha-d2-benzene, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Ethylbenzene-alpha,alpha-D2 |
| CAS | 1861-01-4 |
| Molecular Formula | C8 D2 H8 |
| Quantity | 0.25 g |
| Synonym | Benzene, ethyl-1,1-d2- (6 CI, 7 CI, 8 CI, 9 CI, ACI), Ethyl-1,1-d2-benzene (ACI), (Ethyl-α,α-d2)benzene, 1,1-Dideuterioethylbenzene, Ethylbenzene-d2, Ethylbenzene-α,α-d2, Ethylbenzene-α,α-D2 |
| InChI Formula | InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i2D2 |
| SMILES | [2 H]C([2 H])(C)c1ccccc1 |
| IUPAC Name | 1,1-dideuterioethylbenzene |
| Molecular Weight (g/mol) | 108.1773 |
| Formula Weight | 108.0908 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction