Learn More
Ethyl Hexadecanoate-d31, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic Acid Ethyl Ester-D31 (hexadecanoic-D31) |
| CAS | 1215721-57-5 |
| Molecular Formula | C18 D31 H5 O2 |
| Quantity | 0.25 g |
| Synonym | Ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecanoate, Hexadecanoic-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16-d31 acid ethyl ester, Ethyl hexadecanoate-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16-d31, Hexadecanoic-d31 acid ethyl ester, Palmitic-d31 acid ethyl ester, Ethyl hexadecanoate-d31, Ethyl palmitate-d31, Hexadecanoic Acid Ethyl Ester-D31 (hexadecanoic-D31) |
| InChI Formula | InChI=1 S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3/i1D3,3D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OCC |
| IUPAC Name | ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecanoate |
| Molecular Weight (g/mol) | 315.6683 |
| Formula Weight | 315.4661 g/mol |
| Show More |