Learn More
(±)-Glycidol-1,1,2,3,3-d5, CDN
Product Code. 30312496
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312496
Brand: CDN CDND74830.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Glycidol-d5
CAS 1246819-20-4
Molecular Formula C3 2H5 H O2
Quantity 0.25 g
Synonym Dideuterio-(2,3,3-trideuteriooxiran-2-yl)methanol
InChI Formula InChI=1 S/C3H6O2/c4-1-3-2-5-3/h3-4 H,1-2H2/i1D2,2D2,3 D
SMILES [2 H]C([2 H])(O)C1([2 H])OC1([2 H])[2 H]
IUPAC Name dideuterio-(2,3,3-trideuteriooxiran-2-yl)methanol
Molecular Weight (g/mol) 79.11
Formula Weight 79.0682 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity