Learn More
Hexanenitrile-6,6,6-d3, CDN
Product Code. 30321312
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321312
Brand: CDN CDND69310.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Hexanenitrile-6,6,6-D3
CAS 1219805-03-4
Molecular Formula C6 D3 H8 N
Quantity 0.25 g
Synonym Hexanenitrile-6,6,6-D3, 1-Cyanopentane-5,5,5-d3
InChI Formula InChI=1 S/C6H11N/c1-2-3-4-5-6-7/h2-5H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])CCCCC#N
IUPAC Name 6,6,6-trideuteriohexanenitrile
Molecular Weight (g/mol) 100.1767
Formula Weight 100.108 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity