missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodoacetic Acid-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Iodoacetic Acid-D3 |
| CAS | 1219799-36-6 |
| Molecular Formula | C2 D3 I O2 |
| Quantity | 0.25 g |
| Synonym | Iodoacetic Acid-d3, Iodoacetic-d3 Acid, 2-Iodoacetic-2,2-d2 acid-d, Monoiodoacetic acid-d3, Iodoacetic Acid-D3 |
| InChI Formula | InChI=1 S/C2H3IO2/c3-1-2(4)5/h1H2,(H,4,5)/i1D2/hD |
| SMILES | [2 H]OC(=O)C([2 H])([2 H])I |
| IUPAC Name | deuterio 2,2-dideuterio-2-iodoacetate |
| Molecular Weight (g/mol) | 188.967 |
| Formula Weight | 188.9366 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction