Learn More
Isophorone-d8 (2,4,4,6,6-d5; 3-methyl-d3), CDN
Product Code. 30319986
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319986
Brand: CDN CDND23040.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Isophorone-d8 (2,4,4,6,6-d5; 3-methyl-d3)
CAS 14397-59-2
Molecular Formula C9 D8 H6 O
Quantity 0.25 g
Synonym Isophorone D8 (3-Methyl D3, 2,4,4,6,6 D5), 2-Cyclohexen-1-one-2,4,4,6,6-d5, 5,5-dimethyl-3-(methyl-d3)- (7 CI,8 CI,9 CI)
InChI Formula InChI=1 S/C9H14O/c1-7-4-8(10)6-9(2,3)5-7/h4H,5-6H2,1-3H3/i1D3,4 D,5D2,6D2
SMILES [2 H]C1=C(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(C)(C)C([2 H])([2 H])C1=O
IUPAC Name 2,4,4,6,6-pentadeuterio-5,5-dimethyl-3-(trideuteriomethyl)cyclohex-2-en-1-one
Molecular Weight (g/mol) 146.26
Formula Weight 146.1547 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity