missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenyl-d5-alanine, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | L-Phenylalanine-D5 |
| CAS | 56253-90-8 |
| Molecular Formula | C9 2H5 H6 N O2 |
| Quantity | 0.25 g |
| Synonym | L-Phenylalanine-(phenyl-D5), L-Phenylalanine-2,3,4,5,6-d5, L-Phenyl-d5-alanine (9 CI), L-(2,3,4,5,6-2H5)Phenylalanine, L-Phenyl-2,3,4,5,6-d5-alanine, L-Phenylalanine-d5, [2H5]Phenylalanine, d5-ring-L-Phenylalanine |
| InChI Formula | InChI=1 S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8 H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C[C@H](N)C(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | (2 S)-2-amino-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 170.22 |
| Formula Weight | 170.1104 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction