Learn More
N,N-Dimethyl-4-phenyl-d5-azoaniline, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-(Phenylazo)-N,N-dimethylaniline-D5 (phenyl-D5) |
| CAS | 1398109-08-4 |
| Molecular Formula | C14 D5 H10 N3 |
| Quantity | 0.25 g |
| Synonym | Sudan Yellow-d5, N,N-Dimethyl-4-phenyl-d5-azoaniline, N,N-Dimethyl-4-(2,3,4,5,6-pentadeuteriophenyl)diazenylaniline, 4-(Phenyl-d5-azo)-N,N-dimethylaniline, 4-(Phenylazo)-N,N-dimethylaniline-D5 (phenyl-D5) |
| InChI Formula | InChI=1 S/C14H15N3/c1-17(2)14-10-8-13(9-11-14)16-15-12-6-4-3-5-7-12/h3-11 H,1-2H3/i3D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N=Nc2ccc(cc2)N(C)C)c([2 H])c1[2 H] |
| IUPAC Name | N,N-dimethyl-4-[(2,3,4,5,6-pentadeuteriophenyl)diazenyl]aniline |
| Molecular Weight (g/mol) | 230.32 |
| Formula Weight | 230.158 g/mol |
| Show More |