Learn More
o-Toluidine-4,6-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | o-Toluidine-4,6-D2 |
| CAS | 68408-20-8 |
| Molecular Formula | C7 D2 H7 N |
| Quantity | 0.25 g |
| Synonym | Benzen-2,4-d2-amine, 6-methyl- (9 CI), 6-Methylbenzen-2,4-d2-amine, 4,6-Dideuterio-2-methyl-aniline, o-Toluidine-d2, o-Toluidine-4,6-D2, 2-Toluidine-4,6-d2 |
| InChI Formula | InChI=1 S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5 H,8H2,1H3/i2D,5 D |
| SMILES | [2 H]c1cc([2 H])c(N)c(C)c1 |
| IUPAC Name | 2,4-dideuterio-6-methylaniline |
| Molecular Weight (g/mol) | 109.165 |
| Formula Weight | 109.086 g/mol |
| Show More |