Learn More
Sodium Hexanoate-d11, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Sodium Hexanoate-D11 |
| CAS | 340257-58-1 |
| Molecular Formula | C6 D11 O2 . Na |
| Quantity | 0.25 g |
| Synonym | Hexanoic-d11 acid, sodium salt (9 CI), Hexanoic acid-d11 sodium salt, Hexanoic-D11 acid sodium salt, Caproic-d11 acid sodium salt, Sodium hexanoate-D11, Sodium hexanoate-d11, Sodium caproate-d11 |
| InChI Formula | InChI=1 S/C6H12O2.Na/c1-2-3-4-5-6(7)8;/h2-5H2,1H3,(H,7,8);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)[O-] |
| IUPAC Name | sodium;2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoate |
| Molecular Weight (g/mol) | 149.208 |
| Formula Weight | 149.135 g/mol |
| Show More |