missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene-3-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Toluene-3-D |
| CAS | 4409-85-2 |
| Molecular Formula | C7 D H7 |
| Quantity | 0.25 g |
| Synonym | Benzene-d, 3-methyl- (9 CI, ACI), 3-Methylbenzene-d (ACI), Toluene-m-d (7 CI, 8 CI), m-Deuteriotoluene, 3-Deuteriotoluene, 3-Deuterotoluene, Toluene-3-d, Toluene-3-D |
| InChI Formula | InChI=1 S/C7H8/c1-7-5-3-2-4-6-7/h2-6 H,1H3/i3D |
| SMILES | [2 H]c1cccc(C)c1 |
| IUPAC Name | 1-deuterio-3-methylbenzene |
| Molecular Weight (g/mol) | 93.145 |
| Formula Weight | 93.069 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction