missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-butyl-d27-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Tributylamine-D27 |
| CAS | 1219805-78-3 |
| Molecular Formula | C12 D27 N |
| Quantity | 0.25 g |
| Synonym | 1-Butan-1,1,2,2,3,3,4,4,4-d9-amine, N,N-di(butyl-1,1,2,2,3,3,4,4,4-d9)- (ACI), N,N-Di(butyl-1,1,2,2,3,3,4,4,4-d9)-1-butan-1,1,2,2,3,3,4,4,4-d9-amine (ACI), N,N-Di(butyl-d9)-1-butan-d9-amine, Tributylamine-d27, Tris-(n-butyl-d9)amine, Tri-n-butyl-d27-amine |
| InChI Formula | InChI=1 S/C12H27N/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,4-nonadeuterio-N,N-bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)butan-1-amine |
| Molecular Weight (g/mol) | 212.5158 |
| Formula Weight | 212.3838 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction