Learn More
Motic™ 0.35X C-Mount Camera Adapter
Product Code. 15429254
Change view
Click to view available options
Type:
Camera Adapter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15429254 Camera Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15429254 Supplier Motic™ Supplier No. 1101001904111
  • ONLINE EXCLUSIVE PRICE
    209.00€
    186.04€ / Pack of 1
    Save 22.96€
    10% Off
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This C-mount camera adapter allows Moticams to connect with Motic™ trinocular microscopes.
Compatible with the following cameras:
  • Moticam 2
  • Moticam X
  • Moticam X3
  • Moticam 1080
  • Moticam BTU 10
  • Moticam BTX 8
Specifications
Type Camera Adapter
For Use With (Equipment) Motic BA, AE2000/AE31E, SMZ-161/171 Trinocular Microscopes