missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Benzenediamine-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,3-Phenylenediamine-d4 |
| CAS | 770735-58-5 |
| Molecular Formula | C6 D7 H N2 |
| Quantity | 0.5 g |
| Synonym | 1,3-Benzene-2,4,5,6-d4-di(amine-d2) (9 CI), 1,3-Benzenediamine-d8, m-Phenylenediamine-d8, 1,3-Diaminobenzene-d8, 1,3-Phenylenediamine-d8, 3-Aminoaniline-d8, m-Diaminobenzene-d8, 1,3-Phenylenediamine-D8 |
| InChI Formula | InChI=1 S/C6H8N2/c7-5-2-1-3-6(8)4-5/h1-4 H,7-8H2/i1D,2 D,3 D,4 D/hD3 |
| SMILES | [2 H]Nc1c([2 H])c([2 H])c([2 H])c(N([2 H])[2 H])c1[2 H] |
| IUPAC Name | 1-N,1-N,3-N,2,4,5,6-heptadeuteriobenzene-1,3-diamine |
| Molecular Weight (g/mol) | 115.1843 |
| Formula Weight | 115.1127 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction