Learn More
1,3-Propane-d6-diol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 1,3-Propanediol-d6 |
| CAS | 284474-77-7 |
| Molecular Formula | C3H2D6O2 |
| Quantity | 0.5 g |
| Synonym | 1,3-Dihydroxypropane-d6, 1,3-Propylene Glycol-d6, 1,3-Propylenediol-d6, 2-Deoxyglycerol, Bio-PDO, NSC 65426, PG, Susterra, Trimethylene glycol, Zemea, Zemea propanediol, β-Propylene glycol, ω-Propanediol |
| InChI Formula | InChI=1 S/C3H8O2/c4-2-1-3-5/h4-5 H,1-3H2/i1D2,2D2,3D2 |
| SMILES | OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | propane-d6-1,3-diol |
| Molecular Weight (g/mol) | 82.13 |
| Formula Weight | 82.09 g/mol |
| Show More |