Learn More
1,4-Benzenediamine-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,4-Phenylenediamine-d8 |
| CAS | 45619-88-3 |
| Molecular Formula | C6 D8 N2 |
| Quantity | 0.5 g |
| Synonym | 1,4-Benzene-2,3,5,6-d4-di(amine-d2) (9 CI), 1,4-Phenylenediamine D8, 1,4-Phenylenediamine-d8, Benzene-1,4-diamine-d8, Perdeuterobenzene-1,4-diamine |
| InChI Formula | InChI=1 S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4 H,7-8H2/i1D,2 D,3 D,4 D/hD4 |
| SMILES | [2 H]N([2 H])c1c([2 H])c([2 H])c(N([2 H])[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 1-N,1-N,4-N,4-N,2,3,5,6-octadeuteriobenzene-1,4-diamine |
| Molecular Weight (g/mol) | 116.1904 |
| Formula Weight | 116.119 g/mol |
| Show More |