missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butane-d8-diamine 2HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 1,4-Diaminobutane-D8 Dihydrochloride |
| CAS | 284665-22-1 |
| Molecular Formula | C4 D8 H4 N2 . 2 Cl H |
| Quantity | 0.5 g |
| Synonym | 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine, dihydrochloride (9 CI), 1,4-Butane-d8-diamine dihydrochloride, 1,4-Diaminobutane D8 dihydrochloride, 1,4-Diaminobutane-D8 dihydrochloride, 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine dihydrochloride, 1,4-Butane-d8-diamine d |
| InChI Formula | InChI=1 S/C4H12N2.2 ClH/c5-3-1-2-4-6;;/h1-6H2;2*1 H/i1D2,2D2,3D2,4D2;; |
| SMILES | Cl.Cl.[2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine;dihydrochloride |
| Molecular Weight (g/mol) | 169.123 |
| Formula Weight | 168.104 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction