Learn More
1-Bromo-3-chloro-5-fluorobenzene-d3, CDN
Product Code. 30310993
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310993
Brand: CDN CDND72200.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromo-3-chloro-5-fluorobenzene-D3
CAS 1219805-00-1
Molecular Formula C6 D3 Br Cl F
Quantity 0.5 g
Synonym 1-Bromo-3-chloro-5-fluorobenzene-D3, 1-Fluoro-3-chloro-5-bromobenzene-d3, 3-Chloro-5-fluorobromobenzene-d3
InChI Formula InChI=1 S/C6H3BrClF/c7-4-1-5(8)3-6(9)2-4/h1-3 H/i1D,2 D,3 D
SMILES [2 H]c1c(F)c([2 H])c(Br)c([2 H])c1Cl
IUPAC Name 1-bromo-3-chloro-2,4,6-trideuterio-5-fluorobenzene
Molecular Weight (g/mol) 212.4619
Formula Weight 210.9279 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity