Learn More
2-Chloro-2-methylpropane-2-13C, CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-2-methylpropane-2-13 C |
| CAS | 32488-42-9 |
| Molecular Formula | 13 C C3 H9 Cl |
| Quantity | 0.5 g |
| Synonym | Propane-2-13 C, 2-chloro-2-methyl- (6 CI, 7 CI, 9 CI), 2-Chloro-2-methylpropane-2-13 C, tert-Butyl-13 C chloride, tert-Butyl Chloride-13 C |
| InChI Formula | InChI=1 S/C4H9Cl/c1-4(2,3)5/h1-3H3/i4+1 |
| SMILES | C[13 C](C)(C)Cl |
| IUPAC Name | 2-chloro-2-methyl(213 C)propane |
| Molecular Weight (g/mol) | 93.5599 |
| Formula Weight | 93.0426 g/mol |
| Show More |