missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Methylbutyric-d9 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 2-(Methyl-d3)-butanoic-2,3,3,4,4,4-d6 Acid |
| CAS | 352431-44-8 |
| Molecular Formula | C5 D9 H O2 |
| Quantity | 0.5 g |
| Synonym | 2-Methylbutyric-d9 Acid, α-Methylbutyric-d9 Acid, (±)-2-Methylbutanoic-d9 Acid, (±)-2-Methylbutyric-d9 Acid, (±)-Methylethylacetic-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid, (±)-α-Methylbutyric-d9 Acid |
| InChI Formula | InChI=1 S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/i1D3,2D3,3D2,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,3,4,4,4-hexadeuterio-2-(trideuteriomethyl)butanoic acid |
| Molecular Weight (g/mol) | 111.19 |
| Formula Weight | 111.1246 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction