missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyrrolidinone-3,3,4,4,5,5-d6, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Pyrrolidinone-D6 |
| CAS | 70607-84-0 |
| Molecular Formula | C4 D6 H N O |
| Quantity | 0.5 g |
| Synonym | 2-Pyrrolidinone-3,3,4,4,5,5-d6 (9 CI), Pyrrolidone-d6, Pyrrolidin-2-one-3,3,4,4,5,5-d6, 2-Pyrrolidinone-D6 |
| InChI Formula | InChI=1 S/C4H7NO/c6-4-2-1-3-5-4/h1-3H2,(H,5,6)/i1D2,2D2,3D2 |
| SMILES | [2 H]C1([2 H])NC(=O)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 3,3,4,4,5,5-hexadeuteriopyrrolidin-2-one |
| Molecular Weight (g/mol) | 91.1415 |
| Formula Weight | 91.0904 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction