missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorotoluene-alpha-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Fluorotoluene-alpha-D |
| CAS | 131088-90-9 |
| Molecular Formula | C7 D H6 F |
| Quantity | 0.5 g |
| Synonym | Benzene, 1-fluoro-4-(methyl-d)- (9 CI, ACI), 1-Fluoro-4-(methyl-d)benzene (ACI), 4-Fluorotoluene-α-d1, 4-Fluorotoluene-α-D |
| InChI Formula | InChI=1 S/C7H7F/c1-6-2-4-7(8)5-3-6/h2-5 H,1H3/i1D |
| SMILES | [2 H]Cc1ccc(F)cc1 |
| IUPAC Name | 1-(deuteriomethyl)-4-fluorobenzene |
| Molecular Weight (g/mol) | 111.135 |
| Formula Weight | 111.0595 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction