missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-d3-benzaldehyde, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 4-(Methoxy-d3)benzaldehyde |
| CAS | 342611-04-5 |
| Molecular Formula | C8 D3 H5 O2 |
| Quantity | 0.5 g |
| Synonym | 4-Methoxybenzaldehyde-d3 |
| InChI Formula | InChI=1 S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccc(C=O)cc1 |
| IUPAC Name | 4-(trideuteriomethoxy)benzaldehyde |
| Molecular Weight (g/mol) | 139.17 |
| Formula Weight | 139.0713 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction