missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylonitrile-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Acrylonitrile (D3) |
| CAS | 53807-26-4 |
| Molecular Formula | C3 D3 N |
| Quantity | 0.5 g |
| Synonym | 2-Propenenitrile-2,3,3-d3 (9 CI), Acrylonitrile-d3 (7 CI), Perdeuterioacrylonitrile, Acrylonitrile-D3 |
| InChI Formula | InChI=1 S/C3H3N/c1-2-3-4/h2H,1H2/i1D2,2 D |
| SMILES | [2 H]C(=C([2 H])C#N)[2 H] |
| IUPAC Name | 2,3,3-trideuterioprop-2-enenitrile |
| Molecular Weight (g/mol) | 56.0811 |
| Formula Weight | 56.0454 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction