missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl-d10-amine, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Diethyl-d10-amine |
| CAS | 120092-66-2 |
| Molecular Formula | C4 2H10 H N |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,2-pentadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine |
| Molecular Weight (g/mol) | 83.2 |
| Formula Weight | 83.1519 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction