missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl-1,1,1-d3 Sulfide, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Dimethyl Sulfide-D3 (methyl-D3) |
| CAS | 12-9-4752 |
| Molecular Formula | C2 D3 H3 S |
| Quantity | 0.5 g |
| Synonym | Methane-d3, (methylthio)- (9 CI), (Methyl sulfide)-1,1,1-d3 (7 CI, 8 CI), Methyl perdeuteromethyl sulfide, Methyl trideuteriomethyl sulfide, Trideuteromethyl methyl sulfide, Dimethyl Sulfide-D3 (methyl-D3) |
| InChI Formula | InChI=1 S/C2H6S/c1-3-2/h1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])SC |
| IUPAC Name | trideuterio(methylsulfanyl)methane |
| Molecular Weight (g/mol) | 65.1525 |
| Formula Weight | 65.0379 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction