Learn More
Hexanedinitrile-d8, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Adiponitrile-d8 |
| CAS | 34006-32-1 |
| Molecular Formula | C6 D8 N2 |
| Quantity | 0.5 g |
| Synonym | Hexanedinitrile-d8 (9 CI), Adiponitrile-d8 (8 CI), 2,2,3,3,4,4,5,5-Octadeuteriohexanedinitrile, Hexanedinitrile-D8, 1,4-Dicyanobutane-2,2,3,3,4,4,5,5-d8, Adipic dinitrile-d8, Adipodinitrile-d8, Butanedicarbonitrile-d8 |
| InChI Formula | InChI=1 S/C6H8N2/c7-5-3-1-2-4-6-8/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(C#N)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C#N |
| IUPAC Name | 2,2,3,3,4,4,5,5-octadeuteriohexanedinitrile |
| Molecular Weight (g/mol) | 116.19 |
| Formula Weight | 116.119 g/mol |
| Show More |